Learn More
Fuming Hotplate
SDPSCI_ED
Catalog No. S69026A Shop All Sirchie Finger Print Products
Change view
Click to view available options
For Use With (Application):
For cyanoacrylate development in a fuming chamber
Catalog No. For Use With (Application)
S69026A For cyanoacrylate development in a fuming chamber
1 options
This item is not returnable. View return policy
Catalog No. S69026A Supplier Sirchie Finger Print Supplier No. FHP100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accelerate development of cyanoacrylate by using this new fuming hotplate.
  • Place hotplate into fuming chamber and add 5-10 drops of Omega-Print™ liquid in a disposable, aluminum fuming tray
  • Prints develop in minutes — not hours
  • 110V AC operation
Specifications
Product Type Fuming Hot Plate
For Use With (Application) For cyanoacrylate development in a fuming chamber