Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumitremorgin C, MP Biomedicals™

Catalog No. MP21836832 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21836832 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
UN 346
Catalog No. MP21836832 Supplier MP Biomedicals, Inc Supplier No. 02183683.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UN 346

Fumitremorgin C is extremely effective in reversing resistance to mitoxantrone, doxorubicin, and topotecan in multidrug-selected cell lines

  • Used in breast cancer resistance protein (BCRP; ABCG2) research
  • In Vitro, increases cytotoxicity of anti cancer reagents, and reverses multidrug resistance mediated by BCRP
TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 μg
Chemical Name or Material Fumitremorgin C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.