Learn More
Cayman Chemical Fumonisin B1, 116355-83-0, 1 mg
SDP
Supplier:  Cayman Chemical 625801
Encompass_Preferred
Molecular Formula C34H59NO15, Purity ≥95%, Formula Weight 721.8, Synonyms: FB1; A mycotoxin produced from F. moniliforme that functions as an inhibitor of ceramide synthase (sphingosine N-acyltransferase).
Catalog No. NC9629209
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.