Learn More
Apexbio Technology LLC Fumonisin B1(Synonyms: FB1, Fumonisin-B1, Fusarium Moniliforme toxin B1, Macrofusine, Fusariotoxin B1), 5mg, CAS: 116355-83-0.
SDP
Supplier:  Apexbio Technology LLC A45375
Encompass_Preferred
Fumonisin B1 (CAS 116355-83-0) is a mycotoxin isolated from the fungus Fusarium moniliforme It potently inhibits sphingosine N-acyltransferase (ceramide synthase) an enzyme critical to sphingolipid biosynthesis leading to the intracellular accumulation of sphingoid bases (IC 75 nM) Additionally Fumonisin B1 exhibits inhibitory activity against several protein phosphatases notably PP5 PP2C PP2A PP1 2 and PP2B with IC ranging from approximately 80 M (PP5) to 3000 M (PP2B) This compound is employed in biomedical research to investigate sphingolipid metabolism cell signaling and phosphatase-related cellular processes
Catalog No. 50-000-60804
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.