Learn More
Apexbio Technology LLC PF-04217903(Synonyms: PF04217903, PF 04217903, PF-4217903, PF4217903, MET inhibitor PF-04217903), 25mg, CAS: 956905-27-4.
SDP
Supplier:  Apexbio Technology LLC A509625
Encompass_Preferred
PF-04217903 (CAS 956905-27-4) is a selective ATP-competitive inhibitor targeting the receptor tyrosine kinase c-Met with a reported Ki value of 4 8 nM Aberrant activation or amplification of c-Met signaling contributes significantly to tumor cell proliferation angiogenesis and metastasis PF-04217903 exhibits high selectivity for c-Met over numerous other kinases and inhibits various c-Met mutants including R988C V1092I H1094R and T1010I In cellular studies using lines such as GTL-16 and H1993 PF-04217903 inhibits proliferation and induces apoptosis In vivo it effectively reduces c-Met phosphorylation and tumor growth suggesting potential as a research tool for investigating c-Met-related oncogenic pathways
Catalog No. 50-000-60805
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.