Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fungal beta glucan Recombinant Rabbit Monoclonal Antibody (8201)
GREENER_CHOICE

Catalog No. PIMA533305
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533305 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533305 Supplier Invitrogen™ Supplier No. MA533305
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Reactive with Candida (5 species), Aspergillus (4 species), Penicillium notatum, Saccharomyces cerevisiae, Pichia pastoris, and Mucor hiemalis. Others not tested. Extraction of specimen with 0.4M NaOH required for optimum sensitivity.

Beta-glucans are naturally occurring glucose polymers that are present in abundance in fungi cell walls.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fungal beta glucan
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8201
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.5
Host Species Rabbit
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.