Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fura-2 K+ Salt, Fluorescent Ca2+ Binding Dye, 1 mg

Catalog No. 502706185
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6185 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6185 Supplier Abcam Supplier No. AB1427771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent Ca2²+ binding dye that enables high-affinity calcium detection with excitation at 340/380 nm and emission at 505 nm.

  • Fluorescent Ca²+ binding dye with high affinity (Kd = 225 nM in cytosol)
  • Fluoresces green with increased intensity upon Ca²+ binding
  • Excitation: 340/380 nm
  • Emission: 505 nm
  • Molecular weight: 832 Da
  • Purity: >95%
  • Cell membrane impermeable
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Calcium
Shipping Condition Ambient
Substrate Calcium
Content And Storage Store at -20°C.
Form Solid
Detection Method Fluorescence
Quantity 1 mg
For Use With (Application) Fluorescence detection, Calcium binding
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.