Learn More
AdipoGen Fura-2 pentapotassium salt (1 mg)
SDP
Supplier:  AdipoGen CDXF0013M001
Encompass_Preferred
Chemical. CAS: 113694-64-7. Formula: C29H22N3O14 . 5K. MW: 831.99. Fura-2 pentapotassium salt is a widely used UV-excitable fluorescent calcium indicator. It is a fluorophore chelating probe for Ca2+ used widely in biological systems to analyze intracellular calcium levels. It can be used to detect calcium in cells. A diagnostic shift in the excitation spectrum from 363nm to 335nm indicates FURA-2 chelation to Ca2+. Upon calcium binding, the fluorescent excitation maximum of the indicator undergoes a blue shift from 363 nm (Ca2+-free) to 335 nm (Ca2+-saturated), while the fluorescence emission maximum is relatively unchanged at ~510 nm. Analysis of Ca2+ flux in and out of cells with Fura-2 introduces a powerful spectrographic handle to investigate and elucidate correlated biological events such as neuronal ion channel activation. A major use of Fura-2 is in drug discovery, for measuring intracellular calcium mobilization after activation of GPCRs and calcium channels.
Catalog No. 50-205-4431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.