Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl alcohol, 98%

Catalog No. p-7081115
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1096818 50 g
AAA1096830 250 g
AAA109680B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1096818 Supplier Thermo Scientific Chemicals Supplier No. A1096818
Only null left
Add to Cart

CAS: 98-00-0 | C5H6O2 | 98.101 g/mol

Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO

Specifications

Melting Point -31°C
Density 1.129
Boiling Point 170°C to 171°C
Flash Point 65°C (149°F)
Odor Mild
Refractive Index 1.487
Quantity 50 g
UN Number UN2874
Beilstein 106291
Merck Index 14,4305
Solubility Information Soluble in water.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material Furfuryl alcohol
Show More Show Less
Hazard Category H227-H301+H311-H319-H330-H335-H373
Hazard Statement GHS H Statement
H331-H351-H373-H227-H302-H312-H319-H335
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P262-P264-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P362-P374-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FURFURYL ALCOHOL
EINECSNumber 202-626-1
RTECSNumber LU9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.