Learn More
Sigma Organic Chemistry Furfurylamine | 5G | 617-89-0 | MFCD00003258
Supplier: Sigma Organic Chemistry F200095G

Furfurylamine, 5G
About This Item:
Synonym(s): 2-Aminomethylfuran
Linear Formula: C5H7NO
Molecular Weight: 97.12
CAS Number: 617-89-0
MDL Number: MFCD00003258
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 145-146 C (lit.)
Melting Point: -70 C (lit.)
Density: 1.099 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.490 (lit.)
Storage: room temp
EINECS Number: 210-536-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.