Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furil 98.0+%, TCI America™
SDP

Catalog No. F007825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F007825G 25 g
F00785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F007825G Supplier TCI America Supplier No. F007825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 492-94-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00003241
InChI Key SXPUVBFQXJHYNS-UHFFFAOYSA-N
Synonym furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
PubChem CID 68119
IUPAC Name 1,2-bis(furan-2-yl)ethane-1,2-dione
SMILES C1=COC(=C1)C(=O)C(=O)C2=CC=CO2

Specifications

Melting Point 164°C
Color Yellow
Quantity 25 g
Formula Weight 190.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Furil
RTECSNumber LV0580000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.