Learn More
TARGETMOL CHEMICALS INC Furimazine
SDP
Supplier:  TARGETMOL CHEMICALS INC T153592MG
Encompass_Preferred
Furimazine is an imidazopyrazinone substrate. Furimazine paired with NanoLuc (Nluc) produced 2.5 million-fold brighter luminescence in mammalian cells relative to Oluc-19 with Coelenterazine.
Catalog No. NC2103302
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.