Learn More
New England Biolabs, Inc. Furin – 250 units
SDP
Supplier:  New England Biolabs, Inc. P8077L
Encompass
Furin is a recombinant, ubiquitous subtilisin-like proprotein convertase with a minimal cleavage site of Arg-X-X-Arg. However, the enzyme prefers the site Arg-X-Lys/Arg-Arg.
Catalog No. 50-811-913
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.