Learn More
Pyramex™ Furix™ Safety Eyewear
Catalog No. 19155838 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Lens Tint:
Blue, Mirror
Brown
Clear
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-838 Black Clear
19-155-839 Black Brown
19-155-840 Black Gray
19-155-841 Black Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-838 Supplier Pyramex™ Supplier No. SB8510DT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Wrap-around sunglass style fits all facial sizes
  • Integrated nose piece
  • Stylish straight back temples
  • Lightweight, contemporary frame
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear
    Frame Color Black
    Weight (Metric) 35.6 g
    Lens Material Polycarbonate
    Frame Material Nylon