Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Furobufen | 38873-55-1 | 268.26 g·mol⁻¹ | C16H12O4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY105808100MG

Encompass_Preferred

Furobufen is an anti-inflammatory research compound reported to produce antiarthritic and antipyretic effects and to provide analgesia in inflamed tissue. Supplied for research use only, it is documented with solubility and storage recommendations for laboratory handling.

  • Has CAS number 38873-55-1.
  • Chemical formula C16H12O4 and molecular weight 268.26 g·mol⁻¹.
  • Supplied as a powder and may be prepared in solvent for experiments.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder at -20°C (stable for 3 years) or 4°C (2 years); in solvent at -80°C (2 years) or -20°C (1 year).
  • Intended for research use only; not for diagnostic or therapeutic procedures.

Catalog No. 50-002-43426


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.