Learn More
Medchemexpress LLC Furobufen | 38873-55-1 | MFCD00866025 | 99.9% | 268.26 g·mol⁻¹ | C16H12O4 | 5 MG

Supplier: Medchemexpress LLC HY1058085MG
Furobufen is a small-molecule research compound reported to produce anti-inflammatory, antiarthritic, antipyretic, and analgesic effects in inflamed tissue. It is supplied as a powder for laboratory use and is accompanied by batch-specific certificates and safety documentation; intended for research use only.
- High chemical purity: 99.9%.
- Molecular formula C16H12O4; molecular weight 268.26 g·mol⁻¹.
- Powder form suitable for preparation of stock solutions.
- Recommended storage: powder -20°C (long term), 4°C (short term); in solvent -80°C.
- Accompanied by datasheet, certificate of analysis, and SDS.
- Used in laboratory inflammation and pain studies.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.