Learn More
ATCC Fusarium oxysporum Schlechtendahl (ATCC 48112SPQ) - Freeze-dried fungal culture, strain IMI 141140, research use
Supplier:  ATCC 48112SPQ
Encompass
Fusarium oxysporum Schlechtendahl is supplied as a freeze-dried ampoule containing viable cells (which may include spores and mycelia). This culture (strain designation IMI 141140) is provided for laboratory research use with recommended cultivation conditions for recovery and growth.
  • Freeze-dried ampoule containing viable cells.
  • Strain designation IMI 141140.
  • Recommended media: YM agar or YM broth; potato dextrose agar; potato sucrose agar.
  • Incubation temperature: 24-26 °C.
  • Atmosphere: Aerobic.
  • Intended for research use only; not for therapeutic, diagnostic, or consumable use.
Catalog No. NC3788954