Learn More
Chemyx Inc FUSION 100-X SYRINGE PUMP
Supplier: Chemyx Inc F710X
Fusion 100-X syringe pump is a modular dual-channel infusing only syringe pump designed to handle the majority of precision dosing applications with ease of use and a system value that is unmatched in its class. Additionally, if the application needs more syringes then Fusion 100-X can add an infuse 11-Syringe Holding Rack infuse to support a maximum of 11 syringes. Fusion 100-X infusion two-channel syringe pump ensures pulseless, reproducible infusion pump rates for a wide range of analytical techniques and applications in chemical, pharmaceutical, and biotech R&D.• Holds syringes from 0.5µL to 60 mL• Rates from 0.0001 µL/min to 157 mL/min• Control via touchscreen, USB, RS232, TTL• Free multistep programmable mode included• Glass Touch Screen
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.