Learn More
Cancer Diagnostics, Inc.™ Fusion Micro Scalpel
SDP
Catalog No. 22500459 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Product Type:
Scalpel (Handle Only)
Scalpel Combo Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-500-459 Scalpel (Handle Only)
22-500-460 Scalpel Combo Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-500-459 Supplier Cancer Diagnostics, Inc.™ Supplier No. FUS000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Turns microtome blades into a knife
  • Get more use out of your old microtome blades—simply secure blade into Fusion handle and use as a knife or scalpel
  • Single-use handle and low-profile blades in dispenser pack available separately
  • Combo pack includes 50 handles and 50 blades
Specifications
Product Type Scalpel (Handle Only)