Learn More
Creative Biomart Tagraxofusp
Supplier:  Creative Biomart THP0196
Encompass_Preferred
The product is an IL-3 conjugated truncated diphtheria toxin. It is composed by the catalytic and translocation domains of diphtheria toxin fused via Met-His linker to a full-length human IL-3.
Catalog No. 50-208-4247
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.