Learn More
Chemyx Inc Fusion 200 10-Syringe Holding Rack (syringe not included)
Supplier:  Chemyx Inc 50003
Encompass
Enhance your Fusion 200 two-channel syringe pump system with this precision-machined syringe holder, which allows for up to 10 simultaneous injections. This expansion syringe rack accommodates both plastic and glass syringes with volumes from 1 μL to 10 mL, or four larger syringes of up to 20 mL. Syringes not included.
Catalog No. NC1693136