Learn More
Innovative Research Inc Factor IX (FIX) Deficient Human Plasma, 50ml
Supplier: Innovative Research Inc IFIXDP50ML
Factor IX (FIX) Deficient Human Plasma 50ml - Factor IX (FIX) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor IX so that the remaining activity is less than or equal to 1%.