Learn More
Beckman Coulter FX6100 ROTOR, FIXED ANGLE
Supplier:  Beckman Coulter 369735
Encompass
FX6100 Rotor, Fixed Angle, Bio-Certified, Aluminum, 6 x 100 mL, 10,200 rpm, 11,400 x g
Catalog No. NC1314576
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.