Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579288
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579288 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579288 Supplier Invitrogen™ Supplier No. PA579288
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Rat Cardiac Muscle Tissue, Mouse Cardiac Muscle Tissue, HEPG2 whole cell, HELA whole cell. IHC: mouse cardiac muscle tissue, rat cardiac muscle tissue, human prostatic cancer tissue.

This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na,K-ATPase. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene FXYD1
Gene Accession No. O00168, O08589, Q9Z239
Gene Alias 0610012C17Rik; 1110006M24Rik; FXYD domain containing ion transport regulator 1; FXYD domain-containing ion transport regulator 1; FXYD1; Phospholemman; PLM; Pml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Symbols FXYD1
Host Species Rabbit
Immunogen E.coli-derived human FXYD1 recombinant protein (Position: E21-R92).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5348, 56188, 58971
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.