Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FXYD3 Monoclonal Antibody (OTI4E3)
GREENER_CHOICE

Catalog No. PIMA525938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525938 Supplier Invitrogen™ Supplier No. MA525938
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FXYD3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

FXYD3 belongs to a small family of FXYD-domain containing regulators of Na+/K+ ATPases which share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD, and containing 7 invariant and 6 highly conserved amino acids. This gene encodes a cell membrane protein that may regulate the function of ion-pumps and ion-channels. This gene may also play a role in tumor progression. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXYD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4E3
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FXYD3
Gene Accession No. Q14802
Gene Alias chloride conductance inducer protein Mat-8; FXYD domain containing ion transport regulator 3; FXYD domain-containing ion transport regulator 3; Fxyd3; Mammary tumor 8 kDa protein; Mat8; Phospholemman-like; phospholemman-like protein; Plml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD3
Gene Symbols FXYD3
Host Species Mouse
Immunogen Full length human recombinant protein of FXYD3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.