Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FYTTD1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734422 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4422 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4422 Supplier Proteintech Group Inc Supplier No. 245601AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

FYTD1 required for mRNA export from the nucleus to the cytoplasm. This protein acts as an adapter that uses the BAT1/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. FYTD1 associates with spliced and unspliced mRNAs simultaneously with THOC4.

Specifications

Antigen FYTTD1
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FYTTD1
Gene Accession No. Q96QD9
Gene Alias FYTTD1, Protein 40 2 3, UAP56 interacting factor, UIF
Gene Symbols FYTTD1
Host Species Rabbit
Immunogen FYTTD1 Fusion Protein Ag20051
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84248
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.