Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD3 Polyclonal Antibody

Catalog No. PIPA578242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578242 Supplier Invitrogen™ Supplier No. PA578242

Rabbit Polyclonal Antibody

Positive Control: Synthetic peptide: 1:8000 Predicted Reactivity: Rat (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene FZD3
Gene Accession No. Q61086, Q9NPG1
Gene Alias AU020229; D930050A07Rik; frizzled 3; frizzled 3, seven transmembrane spanning receptor; frizzled class receptor 3; frizzled family receptor 3; frizzled homolog 3; frizzled homolog 3 (Drosophila); frizzled-3; Fz3; Fz-3; FZD3; hFz3; mFz3
Gene Symbols FZD3
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human Frizzled 3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14365, 7976
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.