Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587605 Supplier Invitrogen™ Supplier No. PA587605
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSLFSCEPIT LRMCQDLPYN TTFMPNLLNH YDQQTAALAM EPFHPMVNLD CSRDFRPFLC ALYAPICMEY GRVTLPCRRL CQRAYSECSK LMEMFGVPWP EDMECSRFPD CDEPYPRLVD LNLAGEPTEG APVAVQRDYG FWCPRELKID PDLGYSFLHV RDCSPPCPNM YFRREELSFA RYF; Positive Samples: SH-SY5Y, HeLa, MCF7, Mouse brain, Mouse kidney; Cellular Location: Apical cell membrane, Cell membrane, Cell surface, Membrane, Multi-pass membrane protein.

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FZD3
Gene Accession No. Q61086, Q9NPG1
Gene Alias AU020229; D930050A07Rik; frizzled 3; frizzled 3, seven transmembrane spanning receptor; frizzled class receptor 3; frizzled family receptor 3; frizzled homolog 3; frizzled homolog 3 (Drosophila); frizzled-3; Fz3; Fz-3; FZD3; hFz3; mFz3
Gene Symbols FZD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 23-205 of human FZD3 (NP_0591081).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266715
Target Species Rat, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.