Learn More
Apexbio Technology LLC G-15(Synonyms: G15, GPER antagonist G15, GPER receptor antagonist G15, GPER inhibitor G15, GPER-selective antagonist G-15), 10mg, CAS: 1161002-05-6.
SDP
Supplier:  Apexbio Technology LLC B546910
Encompass_Preferred
G-15 (CAS 1161002-05-6) is a selective antagonist of G protein-coupled receptor 30 (GPR30) with a Ki of approximately 20 nM GPR30 is an integral membrane receptor residing in the endoplasmic reticulum mediating intracellular signaling responses triggered by ligands such as estradiol G-15 specifically inhibits GPR30-mediated pathways without significantly interacting with estrogen receptors ER or ER even at higher concentrations In cellular studies G-15 blocks estrogen- or G-1-induced elevations of intracellular calcium and PI3K activation In vivo research demonstrates that G-15 disrupts estradiol-associated improvements in spatial learning tasks emphasizing its utility in investigating GPR30 function and estrogen signaling mechanisms
Catalog No. 50-223-9971
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.