Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ G 28UCM
Description
G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.
Specifications
Specifications
| Chemical Name or Material | 3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate |
| CAS | 1094451-90-7 |
| Quantity | 10 mg |
| Target | Fatty Acid Synthase Inhibitors |
| Molecular Formula | C24H16O10 |
| Formula Weight | 464.38 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?