Learn More
Abcam G 418 Sulfate, Aminoglycoside Antibiotic Agent, Solid, 1 g
Description
- Molecular Weight: 692.7 Da
- Form: Solid
- Composition: 100% G-418 Disulphate
- Mechanism of Action: Aminoglycoside antibiotic agent; inhibits amino acid incorporation into proteins and the prokaryotic and eukaryotic elongation cycle
- Biological Activity: Potent Delta7-SMN target sequence inducer; exhibits cytotoxic, antibacterial, and antiprotozoal effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Delta7-SMN |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Buffer | 100% G-418 Disulphate |
| Form | Solid |
| Formulation | 100% G-418 Disulphate |
| Quantity | 1 g |
| For Use With (Application) | Antibacterial, antiprotozoal, protein synthesis inhibition, Delta7-SMN target sequence induction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.