Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam G 418 Sulfate, Aminoglycoside Antibiotic Agent, Solid, 5 g

Catalog No. 502583287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3287 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3287 Supplier Abcam Supplier No. AB1442615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminoglycoside antibiotic agent that inhibits protein synthesis and acts as a potent Delta7-SMN target sequence inducer.

  • Molecular Weight: 692.7 Da
  • Form: Solid
  • Composition: 100% G-418 Disulphate
  • Antibiotic Activity: Aminoglycoside antibiotic agent that inhibits amino acid incorporation into proteins and the prokaryotic and eukaryotic elongation cycle
  • Biological Effects: Potent Delta7-SMN target sequence inducer; exhibits cytotoxic, antibacterial, and antiprotozoal effects in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Delta7-SMN
Shipping Condition Ambient
Form Solid
Alternate Part Number AB144261
Buffer 100% G-418 disulphate
Content And Storage Store at -20°C.
Formulation 100% G-418 Disulphate
Quantity 5 g
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.