Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam G 418 Sulfate, Aminoglycoside Antibiotic Agent, Solid, 500 mg

Catalog No. 502583288
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3288 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3288 Supplier Abcam Supplier No. AB144261500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent aminoglycoside antibiotic and Delta7-SMN target sequence inducer that inhibits prokaryotic and eukaryotic protein synthesis.

  • Molecular Weight: 692.7 Da
  • Form: Solid
  • Composition: 100% G-418 Disulphate
  • Antibiotic Activity: Aminoglycoside agent that inhibits amino acid incorporation into proteins and the prokaryotic and eukaryotic elongation cycle
  • Biological Effects: Potent Delta7-SMN target sequence inducer; demonstrates cytotoxic, antibacterial, and antiprotozoal effects in vivo
  • Citation Count: 3

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Delta7-SMN
Buffer 100% G-418 disulphate
Content And Storage Store at -20°C.
Form Solid
Substrate Properties Chemical Substrate
Formulation 100% G-418 Disulphate
Quantity 500 mg
For Use With (Application) Antibacterial, antiprotozoal, protein synthesis inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.