Learn More
Eisco™ g by Free-Fall Apparatus Kit
SCI_ED
Catalog No. S24852 Shop All Eisco Products
Change view
Click to view available options
‘g’ by Free-Fall Apparatus Kit
Catalog No.
S24852 ‘g’ by Free-Fall Apparatus Kit
1 options
Catalog No. S24852 Supplier Eisco™ Supplier No. GFYE14
Add to Cart
Add to Cart
The ‘g’ by Free-Fall Apparatus Kit demonstrates determination of gravitational constant 'g'.
  • Includes wooden base, 1.1 m rod, movable electromagnet, timer
  • Electromagnet powered by timer
  • Supplied with manual
Specifications
Product Type Kit, Free-Fall Apparatus
For Use With (Application) Determination of gravitational constant g
Includes Wooden base, 1.1 m rod, movable electromagnet, timer