Learn More
ACROBiosystems ACROBiosystems Biotinylated Human G-CSF R / CD114 Protein, Avitag™,His Tag
Supplier: ACROBiosystems GCRH82E425UG
Recombinant Protein;25UG;Biotinylated Human G-CSF R, Avitag,His Tag (GCR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 69.2 kDa. The protein migrates as 72-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human G-CSF, Tag Free (Cat. No. GCF-H5214) at 2 μg/mL (100 μL/well) can bind Biotinylated Human G-CSF R, Avitag,His Tag (Cat. No. GCR-H82E4) with a linear range of 0.156-1.25 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.