Learn More
CPC Scientific H-Gly-Pen-Gly-Arg-Gly-Asp-Ser-Pro-Cys-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific FIBN011B
Encompass
Cyclic RGD peptide used in the study of vascular wound response that causes vasodilatation when applied to isolated rat cremaster arterioles.ONE-LETTER SEQUENCE: G-Pen-GRGDSPCA(Pen2 and 9 bridge)MOLECULAR FORMULA: C35H57N13O14S2MOLECULAR WEIGHT:948.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [111844-17-8], RESEARCH AREA: Inflammation
Catalog No. 50-210-9289
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.