Learn More
CPC Scientific H-Gly-Arg-Gly-Asp-Ser-Pro-OH 5MG
SDP
Supplier:  CPC Scientific FIBN014B
Encompass
Peptide used for the affinity purification of fibronectin receptors. It contains the RGD integrin recognition site of the fibronectin cell binding domain. It also inhibits fibronectin binding to platelet sites, and promotes mouse blastocyst attachment and outgrowth, whereas in solutions it acts as a reversible inhibitor of entactin-mediated blastocyst outgrowth.SEQUENCE: H-Gly-Arg-Gly-Asp-Ser-Pro-OHONE-LETTER SEQUENCE: GRGDSPMOLECULAR FORMULA: C22H37N9O10MOLECULAR WEIGHT: 587.59STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [91037-75-1]SYNONYMS: H-Gly-Arg-Gly-Asp-Ser-Pro-OH, GRGDSPRESEARCH AREA: InflammationREFERENCES:R. Pytela et al., Science, 231, 1559 (1986)SKU(s): FIBN-014A, FIBN-014B
Catalog No. 50-210-9295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.