Learn More
PIP G-Tek™ Paradox™ A6 Cut Resistant Glove
GREENER_CHOICE
Catalog No. 17103061 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
17-103-061 Medium
1 options
Catalog No. 17-103-061 Supplier PIP Supplier No. 16361/M
Add to Cart
Add to Cart
Blue Seamless Knit PolyKor™ Blended Glove with Black Foam Nitrile Coated Grip on Palm and Fingers. Blue Reinforced Thumb Crotch - 21 Gauge - A6- Touchscreen Compatible.
  • Seamless construction offers increased comfort and breathability
  • Premium PolyKor™ blended 21G shell is lightweight and provides excellent dexterity, tactile sensitivity, and cut resistance
  • Foam Nitrile coatings are breathable and durable, designed with a cell structure that disperse fluids on contact for an improved grip
  • Touchscreen compatible, allowing users to operate a touch screen device without removing gloves
  • Reinforced thumb crotch provides longer wear life and increased protection
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Qualifies for Oeko-Tex™ Made in Green by using recycled yarn technology
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Compliant with Global Recycling Standard practices
  • Silicone and latex free
Specifications
Construction Seamless Knit
Coating Material Foam Nitrile
Color Blue, Black
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE, PolyKor Fiber
Product Type Cut- and Abrasion-resistant Glove
Size Medium
Gender Unisex
Lining Seamless Lining
Packaging Type Individual Pair Packs
Puncture Rating ANSI/ISEA 105-16 Level 2
Thumb Style Reinforced