Learn More
Protective Industrial Products G-Tek™ PolyKor™ Blended Gloves
Catalog No. 17720157 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-720-157 2X-Large
17-720-152 X-Small
17-720-153 Small
17-720-154 Medium
17-720-155 Large
17-720-156 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-720-157 Supplier Protective Industrial Products Supplier No. 16354/XXL
Add to Cart
Add to Cart
Seamless Knit PolyKor™ Blended Glove with Nitrile Coated Foam Grip on Palm and Fingers
  • Seamless construction offers increased comfort and breathability
  • PolyKor™ shell is lightweight and provides excellent dexterity, tactile sensitivity and cut resistance
  • Nitrile is a synthetic rubber compound that offers excellent puncture, tear and abrasion resistance
  • Foam Nitrile coatings are breathable and durable, designed with a cell structure that disperse fluids on contact for an improved grip
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded hems for easy size identification
  • Washable, resistant to chemicals, water, and ultraviolet light
  • Static Control Property: Static Dissipative
Specifications
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material PolyKor Fiber
Product Type Cut- and Abrasion-resistant Glove
Size 2X-Large
Gender Unisex
Lining Seamless Lining
Packaging Type Bulk
Puncture Rating ANSI/ISEA 105-16 Level 4