Learn More
PIP™ G-Tek™ Seamless Knit Aramid Fiber / Lycra™ Gloves, Nitrile-Coated Smooth Grip
Catalog No. 191303716F Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Blue
Brown
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-130-3716F 2X-Large Green
19-130-3716A X-Small Red
19-130-3716B Small White
19-130-3716C Medium Blue
19-130-3716E X-Large Yellow
19-130-3716D Large Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-3716F Supplier PIP™ Supplier No. 09K1450/XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Medium weight gloves feature nitrile coating on fingertips and palm for secure grip. Ideal for a variety of wet or dry applications.
  • Seamless knit construction provides comfort without sacrificing dexterity
  • Aramid fiber and lycra™ shell provide high tensile strength relative to its weight, is inherently cut-resistant and will not melt, ignite or conduct electricity
  • Nitrile-coated palm and fingertips for secure dry grip and will not absorb liquids into the coating
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded cuff hems for easy size identification
  • Launderable for extended life and to reduce replacement costs
  • Aramid fiber is light, comfortable and provides enduring protection against heat, cuts, lacerations and abrasions
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Nitrile
Color Black, Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Aramid Fiber, Lycra
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:3221
For Use With (Application) material handling, Glass handling operations, fishing industries, sanitation, general maintenance, Woodworking, waste handling and recycling
Glove Cuff Color Green
Glove Cuff Style Elasticated
Grip Design Smooth
Hem Color Green
Model 09-K1450/XXL
Puncture Rating ANSI/ISEA 105-16 Level 1
Series G-Tek™ KEV™ 09-K1450
Length (English) 10.2 in.
Length (Metric) 26 cm