Learn More
G-Biosciences G-Trap™ Immobilized L-Glutamic Acid Column
SCI_ED
Catalog No. S000770
Change view
Click to view available options
Quantity:
5 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000770 5 x 1 mL
S000771 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000770 Supplier G-Biosciences Supplier No. 7862039
Only null left
Add to Cart
Add to Cart
G-Biosciences offers G-Trap™ Immobilized Amino Acids resin in a pre-packed column format.
Immobilized Amino Acids are prepared by coupling amino acid to preactivated 6% cross-linked agarose. The covalent coupling to agarose (resin) is achieved by cross-linking the amino or sulfhydryl group of the amino acid to the preactivated resin.
Specifications
Product Line GeneBLAzer
Product Type Immobilized L-Glutamic Acid
Quantity 5 x 1 mL