Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ GHB-d6 sodium salt Solution, 100 μg/mL in Methanol (as salt), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco G0031ML
Description
GHB, or γ-hydroxybutyrate, is a central nervous system depressant sold under the brand name Xyrem™ and used medically to treat narcolepsy, excessive daytime sleepiness and in rare circumstances, alcoholism. GHB is also used recreationally as an intoxicant and date rape drug. This internal standard is suitable for quantitation of GHB levels in urine, serum, plasma, or oral fluid by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
Specifications
| 67-56-1, 362049-53-4 | |
| MFCD01861535 | |
| 100 μg/mL in methanol (as salt) | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C4HNaO3D6 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 132.12 | |
| 1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction