Learn More
Cayman Chemical G-1, 881639-98-1, 5 mg
SDP
Supplier:  Cayman Chemical 100089335
Encompass_Preferred
Molecular Formula C21H18BrNO3, Purity ≥98%, Formula Weight 412.3; G-1 is a nonsteroidal, high-affinity, selective agonist of GPR30 that binds with a Ki value of 11 nM. Competitive binding studies in endoplasmic reticulum - (ER-) and ER-expressing cells yielded Ki values for estradiol of 0.30 and 0.38 nM, respectively, with no substantial binding of G-1 at 1 M.{14210} The discovery of G-1, a compound that does not bind classical ERs, should facilitate further physiological experiments to define the role of GPR30 in vivo.
Catalog No. NC1042605
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.