Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G10, >98%, Tocris Bioscience™
SDP

Catalog No. 594410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-441-0 Supplier R&D Systems Supplier No. 5944/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Activator of STING-dependent signaling

Activator of STING-dependent signaling; exhibits no direct binding affinity for STING. Stimulates IRF-3-induced transcription and induces IFNα and IFNγ gene expression in human fibroblasts. Exhibits no effect on NF-κB signaling. Blocks viral replication in vitro.

Chemical Identifiers

CAS 702662-50-8
Molecular Formula C21H16ClFN2O3S
Molecular Weight (g/mol) 430.878
InChI Key NAGKYJATVFXZKN-UHFFFAOYSA-N
Synonym sting agonist-1, 4-2-chloro-6-fluorobenzyl-n-furan-2-ylmethyl-3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxamide, 4-2-chloro-6-fluorophenyl methyl-n-2-furanylmethyl-3,4-dihydro-3-oxo-2h-1,4-benzothiazine-6-carboxamide, 4-2-chloro-6-fluorophenyl methyl-n-furan-2-ylmethyl-3-oxo-1,4-benzothiazine-6-carboxamide, 4-2-chloro-6-fluorophenyl methyl-n-furan-2-ylmethyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxamide
PubChem CID 5077622
IUPAC Name 4-[(2-chloro-6-fluorophenyl)methyl]-N-(furan-2-ylmethyl)-3-oxo-1,4-benzothiazine-6-carboxamide
SMILES C1C(=O)N(C2=C(S1)C=CC(=C2)C(=O)NCC3=CC=CO3)CC4=C(C=CC=C4Cl)F

Specifications

Linear Formula C21H16ClFN2O3S
Quantity 10 mg
Solubility Information Soluble to 100mM in DMSO
Formula Weight 430.88
Chemical Name or Material 4-[(2-Chloro-6-fluorophenyl)methyl]-N-(2-furanylmethyl)-3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-car
Grade >98%
Recommended Storage Store at −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.