Learn More
Grainger Back Chamfering Tool
Supplier:  Grainger 16T461
Encompass_Preferred
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for chamfering, facing, and grooving operations. It features a 16mm shank diameter and uses TPGN160308E-AL inserts with a TiAlN coating for wear resistance and heat resistance. The tool has an insert pocket angle of 45 degrees and can chamfer widths from 0.039" to 0.157". It is capable of chamfering steel, stainless steel, cast iron, aluminum and other materials. The tool provides accurate and repeatable chamfers in one pass. With its modular design, it allows for quick insert changes for efficient machining. The Micro 100 Indexable Chamfering Tool delivers superior chamfer quality and tool life.
Catalog No. 50-276-8261
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.