Learn More
Grainger Worm Gear Hose Clamp 5/8 W 7 3/4 ID PK10
Supplier: Grainger 16P232
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear hose clamp with a band width of 7/16 inch. It has a worm screw drive for tightening and can provide up to 450 pounds of clamping force. The clamp has a temperature range of -100 to 300 degrees Fahrenheit. This hose clamp is made in Switzerland and conforms to DIN 3017 standards. The clamp has a screw housing length of 0.59 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.