Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals G3335

Catalog No. AAJ64528MF
Click to view available options
Quantity:
50 mg
100 mg

Catalog No. AAJ64528MF

Supplier: Thermo Scientific Chemicals J64528MF

Only null left
Add to Cart

CAS: 36099-95-3 | C16H19N3O5, C16H19N3O5 | 333.34 g/mol

A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36099-95-3
Molecular Formula C16H19N3O5, C16H19N3O5
Synonym H-Trp-Glu-OH

Specifications

Quantity 50 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Formula Weight 333.34g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
InChI Key PWIQCLSQVQBOQV-UHFFFAOYNA-N
SMILES NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O
IUPAC Name 2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid
Molecular Weight (g/mol) 333.34
Show More Show Less
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.