Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G3BP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E4A2, Proteintech
SDP

Catalog No. 501736909 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6909 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6909 Supplier Proteintech Group Inc Supplier No. 664861IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen

Specifications

Antigen G3BP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E4A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene G3BP1
Gene Accession No. P97855, Q13283
Gene Alias G3BP, G3BP 1, G3BP1, HDH VIII
Gene Symbols G3bp1
Host Species Mouse
Immunogen G3BP1 Fusion Protein ag3728
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100513766, 10146, 171092, 27041
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.