Learn More
Cell Signaling Technology G4S Linker Rabbit mAb, Alexa Fluor 488 Conjugate, Flow Cytometry Application, 100 µl
SDP
Supplier:  Cell Signaling Technology 50515S
Encompass
The G4S Linker Rabbit mAb (Alexa Fluor 488 Conjugate) is a powerful tool developed for flow cytometric analysis in human cells. This antibody specifically targets the G4S linker, enhancing CAR-T cell research effectiveness. Its unique ability to recognize cell surface-expressed CARs differentiates it from competitors, facilitating precise cellular targeting. Ideal for researchers, it is validated and ready to aid in advanced biological studies.
Catalog No. NC2832233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.