Learn More
Epigentek Inc G6PC Polyclonal Antibody, Biotin Conjulated (50 ul)
SDP
Supplier:  Epigentek Inc A62625050
Encompass_Preferred
Biotin Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Glucose-6-phosphatase protein (82-117AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Glucose-6-phosphatase, G-6-Pase, G6Pase, Glucose-6-phosphatase alpha, G6Pase-alpha, G6PC, G6PT.
Catalog No. 50-123-9299
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.